missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane-d12, for NMR, 99.5 atom % D

Product Code. 10511564 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10511564 25 g 25g
10041960 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10511564 Supplier Thermo Scientific Acros Supplier No. 166280250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

CAS: 1735-17-7 | C6H12 | 96.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For NMR
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1735-17-7
Molecular Formula C6H12
Molecular Weight (g/mol) 96.24
MDL Number MFCD00044212
InChI Key XDTMQSROBMDMFD-LBTWDOQPSA-N
Synonym cyclohexane-d12, 2h12 cyclohexane, cyclohexane d, cyclohexane, d12, 2 h?? cyclohexane, cyclohexane-d12, ≥99.6 atom % d, cyclohexane #, cyclohexane-d12, 99.5 atom % d
PubChem CID 123129
IUPAC Name 1,1,2,2,3,3,4,4,5,5,6,6-dodecadeuteriocyclohexane
SMILES [2H]C1([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C([2H])([2H])C1([2H])[2H]

Specifications

Physical Form Liquid
Color Colorless
Density 0.8900g/mL
Deuterium (D) 99.4% min.
Flash Point −18°C
Refractive Index 1.421
Specific Gravity 0.89
Quantity 25 g
Boiling Point 78.0°C
Linear Formula C6D12
Formula Weight 96.26
Grade NMR
Chemical Name or Material Cyclohexane-d12, 99.5 atom%D
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexane-d12
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • ACUTE AQUATIC Acute 1
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • H400-Very toxic to aquatic life.
  • H410-Very toxic to aquatic life with long lasting effects.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.