missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, Environmental Grade, 99.7+%

Produktkod. 11365307
Change view
Klicka för att se tillgängliga alternativ
Quantity:
4 x 4 L
Förpackningsstorlek:
16 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Produktkod. Quantity unitSize
11365307 4 x 4 L 16 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Denna artikel kan inte returneras. Se returpolicy
Produktkod. 11365307 Leverantör Thermo Scientific Alfa Aesar Leverantörsnummer 040975.M6
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Se alternativa produkter

Denna artikel kan inte returneras. Se returpolicy

Suitable for pesticide residue, environmental and GC analysis | CAS: 110-82-7 | C6H12 | 84.16 g/mol

Used in environmental and GC analysis, for industrial production of adipic acid and caprolactam, which is a precursor of nylon, used as a solvent, and to make nylon and paint remover.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Specifikationer

Melting Point 6.5°C
Color Colorless
Density 0.779 g/mL
Quantity 4 x 4 L
Flash Point -18°C (-1°F)
Odor Pungent
Physical Form Liquid
Refractive Index 1.426
UN Number UN1145
Beilstein 1900225
Merck Index 14,2723
Boiling Point 80°C to 81°C
Solubility Information Immiscible in water and soluble in ether,alcohol and acetone. Miscible with olive oil.
Formula Weight 84.16
Percent Purity ≥99.7%
Grade Environmental
Chemical Name or Material Cyclohexane
Visa mer Visa mindre

RUO – Research Use Only

Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.