missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexane, for pesticide residue analysis

Product Code. 10571911 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Unit Size:
1 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10571911 1 L Glass Bottle 1 L
10449222 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10571911 Supplier Thermo Scientific Acros Supplier No. 326590010
  • ONLINE EXCLUSIVE PRICE
    71.10 €
    65.40 € / 1 L
    Save 5.70 €
    8% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 110-82-7 | C6H12 | 84.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

For residue analysis
TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 110-82-7
Molecular Formula C6H12
Molecular Weight (g/mol) 84.16
MDL Number MFCD00003814
InChI Key XDTMQSROBMDMFD-UHFFFAOYSA-N
Synonym hexamethylene, hexahydrobenzene, hexanaphthene, cyclohexan, cykloheksan, cicloesano, cyclohexaan, benzene, hexahydro, benzenehexahydride, polycyclohexane
PubChem CID 8078
ChEBI CHEBI:29005
IUPAC Name cyclohexane
SMILES C1CCCCC1

Tekniske data

Melting Point 6.5°C
Density 0.7700g/mL
Quantity 1 L
Flash Point −18°C
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.4250 to 1.4270
Residue after Evaporation 2ppm max.
Specific Gravity 0.77
Linear Formula C6H12
Viscosity 0.94 mPa.s (20°C)
Water 0.02% max. (coulometric)
Beilstein 05, 20
Merck Index 15, 2716
Boiling Point 81.0°C
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in methanol (57 g/100 mL at 20°C), ethanol, , diethyl ether, acetone, benzene, ccl4.
Formula Weight 84.15
Grade Pesticide
Chemical Name or Material Cyclohexane, ECD Tested, For Pesticide Analysis
Vis mere Vis mindre
compliance-icons compliance-icons compliance-icons compliance-icons
Produktidentifikator
  • Cyclohexane
Signalord
  • Danger
Farekategori
  • Flammable liquid Category 2
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
  • ACUTE AQUATIC Acute 1
  • LONG-TERM AQUATIC HAZARD Chronic 1
Faresætning
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H336-May cause drowsiness or dizziness.
  • H410-Very toxic to aquatic life with long lasting effects.
Sikkerhedssætning
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.