missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanecarbonyl Chloride, TRC

Product Code. 30354074
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354074 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30354074 Supplier Toronto Research Chemicals Supplier No. TRCC98747510G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclohexanecarbonyl Chloride
CAS 2719-27-9
Molecular Formula C7H11ClO
Quantity 10 g
Synonym (Chlorocarbonyl)cyclohexane, Cyclohexanecarboxylic Acid Chloride, Cyclohexanoic Acid Chloride, Cyclohexanoyl Chloride, Cyclohexylcarbonyl Chloride, Hexahydrobenzoyl Chloride, NSC 85181
InChI Formula InChI=1S/C7H11ClO/c8-7(9)6-4-2-1-3-5-6/h6H,1-5H2
SMILES ClC(=O)C1CCCCC1
IUPAC Name cyclohexanecarbonyl chloride
Molecular Weight (g/mol) 146.61
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.