missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanepropionic acid, 99%, Thermo Scientific Chemicals

Catalog Number p-96594
65.26 EUR valid until 2024-03-29
Use promo code "21615" to get your promotional price.


Click to view available options
Quantity:
50 g
250 g
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
701-97-3
C9H16O2
156.23
MFCD00001527
HJZLEGIHUQOJBA-UHFFFAOYSA-N
cyclohexanepropionic acid, 3-cyclohexylpropionic acid, cyclohexanepropanoic acid, 3-cyclohexanepropionic acid, cyclohexane propionic acid, unii-u36w9hnv6d, 3-cyclohexyl propionic acid, 3-cyclohexyl-propionic acid, u36w9hnv6d, cyclohexyl propionic acid
69702
3-cyclohexylpropanoic acid
OC(=O)CCC1CCCCC1
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21615” to get your promotional price
701-97-3
C9H16O2
156.23
MFCD00001527
HJZLEGIHUQOJBA-UHFFFAOYSA-N
cyclohexanepropionic acid, 3-cyclohexylpropionic acid, cyclohexanepropanoic acid, 3-cyclohexanepropionic acid, cyclohexane propionic acid, unii-u36w9hnv6d, 3-cyclohexyl propionic acid, 3-cyclohexyl-propionic acid, u36w9hnv6d, cyclohexyl propionic acid
69702
3-cyclohexylpropanoic acid
OC(=O)CCC1CCCCC1

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 14.0°C to 17.0°C
Color Colorless to Yellow
Density 0.9980g/mL
Boiling Point 275.7°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula C6H11CH2CH2CO2H
Refractive Index 1.4630 to 1.4650
Quantity 50 g
Beilstein 09, 82
Specific Gravity 0.998
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in organic solvents
Formula Weight 156.22
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cyclohexanepropionic acid, 99%
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'