missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanol, Pure, Fisher Chemical™

Product Code. 10131900 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
500 mL
2.5 L
Unit Size:
2.5 L
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10131900 500 mL 500mL
10439170 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10131900 Supplier Fisher Chemical Supplier No. C/9000/PB08
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 108-93-0
MDL Number 3855

Specifications

Melting Point 23°C
pH 6.5
Boiling Point 161°C
Packaging Plastic Coated Glass bottle
Molecular Formula C6H12O
Quantity 500 mL
Formula Weight 100.16g/mol
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.