missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone oxime, 97%

Product Code. 11428677
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428677 500 g 500g
11418677 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11428677 Supplier Thermo Scientific Alfa Aesar Supplier No. A19820.36
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 100-64-1 | C6H11NO | 113.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 100-64-1
Molecular Formula C6H11NO
Molecular Weight (g/mol) 113.16
MDL Number MFCD00001660
InChI Key VEZUQRBDRNJBJY-UHFFFAOYSA-N
Synonym cyclohexanone oxime, cyclohexanone, oxime, antioxidant d, hydroxyimino cyclohexane, unii-2u60l00cgf, ccris 1383, dsstox_cid_1842, dsstox_rid_76361, dsstox_gsid_21842, cyclohexanoneoxime
PubChem CID 7517
IUPAC Name N-cyclohexylidenehydroxylamine
SMILES ON=C1CCCCC1

Specifications

Melting Point 87°C to 91°C
Boiling Point 206°C to 210°C
Flash Point 103°C (217°F)
Quantity 500 g
Beilstein 1616769
Formula Weight 113.16
Percent Purity 97%
Chemical Name or Material Cyclohexanone oxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.