missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexanone, TRC

Product Code. 30356134
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30356134 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30356134 Supplier Toronto Research Chemicals Supplier No. TRCC98798010G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclohexanone
CAS 108-94-1
Purity Grade Notes GC
Molecular Formula C6 H10 O
Quantity 10 g
Synonym Anon, Anone, C 0489, Hexanon, Hytrol O, NSC 5711, Nadone, Pimelic ketone, Pimelin ketone, Sextone
InChI Formula InChI=1S/C6H10O/c7-6-4-2-1-3-5-6/h1-5H2
SMILES O=C1CCCCC1
IUPAC Name cyclohexanone
Molecular Weight (g/mol) 98.14
Formula Weight 98.0732
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.