missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexene, 99+%, Extra Pure, SLR, Fisher Chemical™

Product Code. 11448153 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
Unit Size:
250mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11448153 250 mL 250mL
10479550 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448153 Supplier Fisher Chemical Supplier No. C/9080/07
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 110-83-8
Molecular Formula C6H10
MDL Number 1539

Specifications

Melting Point -104°C
Color Colorless
pH 7 to 8
Boiling Point 83°C
Packaging Amber glass bottle
Quantity 250 mL
Vapor Density 2.8
Formula Weight 82.13g/mol
Vapor Pressure 94mbar at 20°C
Physical Form Liquid
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexene
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P264-Wash thoroughly after handling.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.