missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexene, 99% stab.

Product Code. 11430163
Change view
Click to view available options
Quantity:
100 mL
500 mL
2500 mL
Unit Size:
100mL
2500mL
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430163 2500 mL 2500mL
11334539 100 mL 100mL
11420163 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11430163 Supplier Thermo Scientific Alfa Aesar Supplier No. A11359.0F
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

CAS: 110-83-8 | C6H10 | 82.146 g/mol

Cyclohexene is used as a solvent. It is employed as a precursor to caprolactam, adipic acid, maleic acid, dicyclohexyladipate and cyclohexeneoxide. It is used as an intermediate for the production of cyclohexanol and cyclohexeneoxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexene is used as a solvent. It is employed as a precursor to caprolactam, adipic acid, maleic acid, dicyclohexyladipate and cyclohexeneoxide. It is used as an intermediate for the production of cyclohexanol and cyclohexeneoxide.

Solubility
Immiscible with cold water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110-83-8
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00001539
InChI Key HGCIXCUEYOPUTN-UHFFFAOYSA-N
Synonym tetrahydrobenzene, cyclohex-1-ene, benzene tetrahydride, 1,2,3,4-tetrahydrobenzene, benzenetetrahydride, hexanaphthylene, 1-cyclohexene, cykloheksen, benzene, tetrahydro, 3,4,5,6-tetrahydrobenzene
PubChem CID 8079
ChEBI CHEBI:36404
IUPAC Name cyclohexene
SMILES C1CCC=CC1

Specifications

Melting Point -104°C
Density 0.811
Boiling Point 83°C to 85°C
Flash Point −20°C (−4°F)
Odor Petrol-like
Refractive Index 1.446
Quantity 2500 mL
UN Number UN2256
Beilstein 906737
Merck Index 14,2727
Solubility Information Immiscible with cold water.
Formula Weight 82.15
Percent Purity 99%
Chemical Name or Material Cyclohexene
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Cyclohexene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P331-Do NOT induce vomiting.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contain: Cyclohexene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.