missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexene, TRC

Product Code. 30357268
Change view
Click to view available options
Quantity:
10 mL
Unit Size:
10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30357268 10 mL 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357268 Supplier Toronto Research Chemicals Supplier No. TRCC98751310ML
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Cyclohexene
CAS 110-83-8
Purity Grade Notes GC
Molecular Formula C6H10
Quantity 10 mL
Synonym 1,2,3,4-Tetrahydrobenzene, Benzene tetrahydride, Benzene, tetrahydro-, Cyclohex-1-ene, NSC 24835, Tetrahydrobenzene
InChI Formula InChI=1S/C6H10/c1-2-4-6-5-3-1/h1-2H,3-6H2
SMILES C1CCC=CC1
IUPAC Name cyclohexene
Molecular Weight (g/mol) 82.14
Formula Weight 82.0783
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.