missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl iodide, 98%, stabilized

Product Code. 10358400 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
25mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10358400 25 mL Glass bottle 25mL
10157353 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. Se returpolicy
Artikelnummer. 10358400 Leverandør Thermo Scientific Acros Leverandørnr. 218490250
 Restricted Item
Estimated Shipment: 29-10-2026
Læg i indkøbskurv
Læg i indkøbskurv
Request Bulk or Custom Format

missing translation for 'validMainframeMsg'

Denne vare kan ikke returneres. Se returpolicy

CAS: 626-62-0 | C6H11I | 210.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiske identifikatorer

CAS 626-62-0
Molecular Formula C6H11I
Molecular Weight (g/mol) 210.06
MDL Number MFCD00003826
InChI Key FUCOMWZKWIEKRK-UHFFFAOYSA-N
Synonym cyclohexane, iodo, cyclohexyl iodide, cyclohexyliodide, iodo-cyclohexane, 1-iodocyclohexane, heptynyltrimethylsilane, iodocyclohexane, acmc-1att4, c-c6h11i
PubChem CID 12289
IUPAC Name iodocyclohexane
SMILES IC1CCCCC1

Tekniske data

Color Brown
Density 1.6200g/mL
Boiling Point 80.0°C to 81.0°C (20.0 mmHg)
Flash Point 71°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H11I
Refractive Index 1.5470 to 1.5490
Quantity 25 mL
Beilstein 05, 25
Specific Gravity 1.62
Formula Weight 210.06
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclohexyl iodide
Vis mere Vis mindre
compliance-icons
Produktidentifikator
  • Cyclohexyl iodide
Signalord
  • Warning
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Faresætning
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Sikkerhedssætning
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Produkttitel
Vælg et problem

Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.