missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ Cyclohexyl isocyanate, 98%

Product Code. 11456410
Change view
Click to view available options
Quantity:
25g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11456410 25g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11456410 Supplier Alfa Aesar™ Supplier No. B23059.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 3173-53-3
Boiling Point 168°C to 170°C
Flash Point 48°C (118°F)
Odor Pungent
Molecular Formula C7H11NO
MDL Number MFCD00003840
Quantity 25g
UN Number UN2488
Beilstein 507983
Synonym cyclohexyl isocyanate, cyclohexylisocyanate, cyclohexane, isocyanato, isocyanato-cyclohexane, isocyanic acid, cyclohexyl ester, unii-t699595blw, isocyanic acid cyclohexyl ester, chi, cyclohexanisocyanate, cyclohexyl-isocyanate
InChI Key KQWGXHWJMSMDJJ-UHFFFAOYSA-N
SMILES O=C=NC1CCCCC1
IUPAC Name isocyanatocyclohexane
Molecular Weight (g/mol) 125.17
PubChem CID 18502
ChEBI CHEBI:136604
Formula Weight 125.17
Percent Purity 98%
Chemical Name or Material Cyclohexyl isocyanate
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.