missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl isocyanide, 99%

Product Code. 10091750 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10091750 1 g Glass bottle 1g
10558873 5 g Glass bottle 5g
10015773 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10091750 Supplier Thermo Scientific Acros Supplier No. 158400010
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
This item is not returnable. View return policy

CAS: 931-53-3 | C7H11N | 109.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 931-53-3
Molecular Formula C7H11N
Molecular Weight (g/mol) 109.17
MDL Number MFCD00003839
InChI Key XYZMOVWWVXBHDP-UHFFFAOYSA-N
Synonym cyclohexyl isocyanide, cyclohexane, isocyano, cyclohexylisocyanide, cyclohexaneisonitrile, cyclohexyl isonitrile, n-cyclohexylisocyanide, cyi, cyclohexylisonitrile, isocyano-cyclohexane, cyclohexyliminocarbene
PubChem CID 79129
ChEBI CHEBI:17966
IUPAC Name isocyanocyclohexane
SMILES [C-]#[N+]C1CCCCC1

Specifications

Color Colorless to Brown
Density 0.8900g/mL
Boiling Point 173.0°C to 176.0°C
Flash Point 77°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H11NC
Refractive Index 1.4490 to 1.4520
Quantity 1 g
Fieser 06,152
Specific Gravity 0.89
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in inert organic solvents
Formula Weight 109.17
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cyclohexyl isocyanide
Show More Show Less
compliance-icons
Product Identifier
  • Cyclohexyl isocyanide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.