missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl isothiocyanate, 98%

Product Code. 11437427
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11437427

Brand: Thermo Scientific Alfa Aesar A15585.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1122-82-3 | C7H11NS | 141.232 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1122-82-3
Molecular Formula C7H11NS
Molecular Weight (g/mol) 141.232
MDL Number MFCD00003841
InChI Key MZSJGCPBOVTKHR-UHFFFAOYSA-N
Synonym cyclohexyl isothiocyanate, cyclohexane, isothiocyanato, isothiocyanocyclohexane, cyclohexyl-isothiocyanat, isothiocyanic acid, cyclohexyl ester, cyclohexylisothiocyanate, cyclohexyl isothiocyanate, isothiocyanato, cyclohexyl-isothiocyanat german, isothiocyanato-cyclohexane, isothiocyanic acid cyclohexyl ester
PubChem CID 14289
IUPAC Name isothiocyanatocyclohexane
SMILES C1CCC(CC1)N=C=S

Specifications

Density 1.033g/mL
Boiling Point 219°C
Odor Pungent
Refractive Index 1.5380
Quantity 25 g
UN Number UN2922
Beilstein 774525
Sensitivity Moisture Sensitive
Formula Weight 141.24
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclohexyl isothiocyanate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclohexyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Cyclohexyl isothiocyanate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.