missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl methacrylate, 97%, stab. with ca 50ppm 4-methoxyphenol, Thermo Scientific Chemicals

Product Code. 11332916
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11332916 100 g 100g
11322916 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11332916 Supplier Thermo Scientific Alfa Aesar Supplier No. L10634.22
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 101-43-9
Molecular Formula C10H16O2
Molecular Weight (g/mol) 168.24
MDL Number MFCD00014292
InChI Key OIWOHHBRDFKZNC-UHFFFAOYSA-N
Synonym cyclohexyl methacrylate, methacrylic acid, cyclohexyl ester, 2-propenoic acid, 2-methyl-, cyclohexyl ester, unii-5l9uuv9t6q, 2-methyl-2-propenoic acid cyclohexyl ester, 5l9uuv9t6q, methacrylic acid cyclohexyl ester, ageflex chma, c-hma, cyclo hexyl methacrylate
PubChem CID 7561
IUPAC Name cyclohexyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OC1CCCCC1

Specifications

Density 0.97
Boiling Point 72°C to 73°C (5 mmHg)
Flash Point 82°C (179°F)
Refractive Index 1.4577
Quantity 100 g
Beilstein 2045235
Formula Weight 168.24
Percent Purity 97%
Chemical Name or Material Cyclohexyl methacrylate, stabilized with ≈50ppm 4-methoxyphenol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.