missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl methyl sulfide, 97%

Product Code. 11362276
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362276 5 g 5g
11372276 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11362276 Supplier Thermo Scientific Alfa Aesar Supplier No. L03712.06
 Restricted Item
Estimated Shipment: 10-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 7133-37-1 | C7H14S | 130.25 g/mol

Cyclohexyl methyl sulfide is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclohexyl methyl sulfide is used as a pharmaceutical intermediate.

Solubility
Soluble in water 173.5 mg/L @ 25°C, alcohol.

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7133-37-1
Molecular Formula C7H14S
Molecular Weight (g/mol) 130.25
MDL Number MFCD00039460
InChI Key QQBIOCGHCKNYGP-UHFFFAOYSA-N
Synonym cyclohexyl methyl sulfide, methylthio cyclohexane, sulfide, cyclohexyl methyl, cyclohexane, methylthio, methylthiocyclohexane, methylcyclohexylsulfide, cyclohexylmethylsulphide, cyclohexyl methylsulfide, methyl cyclohexyl sulfide, methylsulfanyl cyclohexane
PubChem CID 81553
SMILES CSC1CCCCC1

Specifications

Melting Point -80°C
Density 0.944
Boiling Point 89°C to 90°C (35 mmHg)
Flash Point 61°C (142°F)
Odor Unpleasant
Refractive Index 1.494
Quantity 5 g
UN Number UN3334
Beilstein 1919309
Solubility Information Soluble in water 173.5mg/L @ 25°C,alcohol.
IUPAC Name (methylsulfanyl)cyclohexane
Formula Weight 130.25
Percent Purity 97%
Chemical Name or Material Cyclohexyl methyl sulfide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.