missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyl Phenyl Ketone, TRC

Product Code. 30354606
Change view
Click to view available options
Quantity:
250 mg
500 mg
2.5 g
Unit Size:
2.5g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30354606 500 mg 500mg
30362776 2.5 g 2.5g
30368368 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30354606 Supplier Toronto Research Chemicals Supplier No. TRCC992145500MG
 Restricted Item
Estimated Shipment: 04-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 712-50-5
Molecular Formula C13H16O
Molecular Weight (g/mol) 188.26
Synonym Cyclohexylphenylmethanone, Benzoyl cyclohexane
IUPAC Name cyclohexyl(phenyl)methanone
SMILES O=C(C1CCCCC1)c2ccccc2

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C13H16O/c14-13(11-7-3-1-4-8-11)12-9-5-2-6-10-12/h1,3-4,7-8,12H,2,5-6,9-10H2
Formula Weight 188.1201
Chemical Name or Material Cyclohexyl Phenyl Ketone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.