Learn More
Cyclohexylamine, 98+%
Description
Cyclohexylamine is a useful as an intermediate in organic synthesis. It acts as a precursor to prepare pendant-armed dialdehydes, bromohexine HCl and sulfenamide-based reagents. It is widely used in the production of vulcanization accelerators like N-cyclohexyl-2-benzothiazole sulfonamide (CBS) and artificial sweeteners (cyclamates). As a corrosion inhibitor, it is used in the treatment of boiler water and prevents corrosion of metal surfaces that arise due to dissolved carbon dioxide. It is involved as a solvent in the synthesis of an inorganic-organic hybrid semiconductor, ZnS/CHA (CHA = cyclohexylamine) nanocomposites.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Cyclohexylamine is a useful as an intermediate in organic synthesis. It acts as a precursor to prepare pendant-armed dialdehydes, bromohexine HCl and sulfenamide-based reagents. It is widely used in the production of vulcanization accelerators like N-cyclohexyl-2-benzothiazole sulfonamide (CBS) and artificial sweeteners (cyclamates). As a corrosion inhibitor, it is used in the treatment of boiler water and prevents corrosion of metal surfaces that arise due to dissolved carbon dioxide. It is involved as a solvent in the synthesis of an inorganic-organic hybrid semiconductor, ZnS/CHA (CHA = cyclohexylamine) nanocomposites.
Solubility
Miscible with water, ethanol, ethers, acetone, alcohol and ketones.
Notes
Air and moisture sensitive. Incompatible with strong oxidizing agents, carbon dioxide, sodium hypochlorite, organic acids, mineral acids and peroxides.
Specifications
Specifications
| Melting Point | -18°C |
| Density | 0.868 |
| Boiling Point | 133°C to 134°C |
| Flash Point | 27°C (81°F) |
| Refractive Index | 1.4585 |
| Quantity | 2500 mL |
| UN Number | UN2357 |
| Beilstein | 471175 |
| Sensitivity | Air Sensitive |
| Merck Index | 14,2729 |
| Show More |
Safety and Handling
- Danger
- Acute toxicity Category 3
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- Reproductive toxicity Category 2
- Skin corrosion/irritation Category 1 B
- H302-Harmful if swallowed.
- H311-Toxic in contact with skin.
- H314-Causes severe skin burns and eye damage.
- H318-Causes serious eye damage.
- H361f-Suspected of damaging fertility.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.