missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylamine, 99%

Product Code. 10786131 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mL
1 L
2.5 L
10 L
Packaging:
Glass bottle
Plastic drum
Unit Size:
1 L
10 L
2.5 L
250mL
This item is not returnable. View return policy

Product Code. 10786131

Brand: Thermo Scientific Acros 111280010

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 108-91-8 | C6H13N | 99.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Kemiska identifierare

CAS 108-91-8
Molecular Formula C6H13N
Molecular Weight (g/mol) 99.18
InChI Key PAFZNILMFXTMIY-UHFFFAOYSA-N
Synonym cyclohexylamine, aminocyclohexane, hexahydroaniline, hexahydrobenzenamine, aminohexahydrobenzene, cyclohexyl amine, 1-cyclohexylamine, 1-aminocyclohexane, aniline, hexahydro, benzenamine, hexahydro
PubChem CID 7965
ChEBI CHEBI:15773
IUPAC Name cyclohexanamine
SMILES C1CCC(CC1)N

Specifikationer

Melting Point -17.0°C
Density 0.8665g/mL
Boiling Point 133.0°C to 134.0°C
Flash Point 27°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H11NH2
Refractive Index 1.4580 to 1.4600
Quantity 1 L
Beilstein 12,5
Merck Index 15,2722
Specific Gravity 0.8665
Solubility Information Solubility in water: solublee. Other solubilities: miscible with most common organic solvents
Viscosity 2.4 mPa.s (20°C)
Formula Weight 99.18
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material Cyclohexylamine
Visa mer Visa mindre
compliance-icons compliance-icons compliance-icons compliance-icons
Produktbeteckning
  • Cyclohexylamine
Signalord
  • Danger
Riskkategorier
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Reproductive toxicity Category 2
  • Skin corrosion/irritation Category 1 B
Faroangivelser
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H361f-Suspected of damaging fertility.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H226-Flammable liquid and vapour.
Skyddsangivelse
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Korrigering av produktinnehåll

Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.

Produkttitel

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.

Tack! Din feedback har skickats. Fisher Scientific arbetar alltid för att förbättra vårt innehåll åt dig. Vi uppskattar din feedback.