missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylamine hydrobromide, 98%

Product Code. 11395179
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11395179 100 g 100g
11305189 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11395179 Supplier Thermo Scientific Alfa Aesar Supplier No. A18439.22
 Restricted Item
Estimated Shipment: 30-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26227-54-3 | C6H14BrN | 180.089 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26227-54-3
Molecular Formula C6H14BrN
Molecular Weight (g/mol) 180.089
MDL Number MFCD00034986
InChI Key QOHWJRRXQPGIQW-UHFFFAOYSA-N
Synonym cyclohexylamine hydrobromide, cyclohexanamine hydrobromide, cyclohexylammonium bromide, cyclohexanamine, hydrobromide, cyclohexanaminium bromide, cyclohexyl amine hydrobromide, acmc-1ccru, cyclohexylamine hbr, ksc268i9n, cyclohexamine, hydrobromide salt
PubChem CID 117303
IUPAC Name cyclohexanamine;hydrobromide
SMILES C1CCC(CC1)N.Br

Specifications

Melting Point 197°C to 201°C
Quantity 100 g
Beilstein 3908098
Sensitivity Hygroscopic
Formula Weight 180.09
Percent Purity 98%
Chemical Name or Material Cyclohexylamine hydrobromide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.