missing translation for 'onlineSavingsMsg'
Learn More

[(Cyclohexylammonium)methyl]trifluoroborate internal salt, 95%, Thermo Scientific™

Product Code. 15458615
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15458615 250 mg 250mg
15448615 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15458615 Supplier Thermo Scientific Alfa Aesar Supplier No. H33741.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 888711-52-2
Molecular Formula C7H14BF3KN
Molecular Weight (g/mol) 219.1
MDL Number MFCD20134171
InChI Key GYUWPHIATXXXJT-UHFFFAOYSA-N
Synonym potassium cyclohexylamino methyl trifluoroborate, potassium n-cyclohexyl-aminomethyltrifluoroborate, potassium cyclohexylamino methyl trifluoroboranuide, pubchem11576, c7h14bf3n.k, potassium cyclohexylamino methyl trifluoro borate 1-, borate 1-, cyclohexylamino methyl trifluoro-, potassium, t-4-9ci
PubChem CID 45588140
IUPAC Name potassium;(cyclohexylamino)methyl-trifluoroboranuide
SMILES [B-](CNC1CCCCC1)(F)(F)F.[K+]

Specifications

Quantity 250 mg
Formula Weight 181.01
Percent Purity 95%
Chemical Name or Material [(Cyclohexylammonium)methyl]trifluoroborate internal salt
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.