missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylbenzene, 98%

Product Code. 10000870 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
500 mL
Packaging:
Glass bottle
Unit Size:
100mL
500mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10000870 500 mL Glass bottle 500mL
10530191 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10000870 Supplier Thermo Scientific Acros Supplier No. 111295000
 Restricted Item
Estimated Shipment: 10-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 827-52-1 | C12H16 | 160.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 827-52-1
Molecular Formula C12H16
Molecular Weight (g/mol) 160.26
MDL Number MFCD00001451
InChI Key IGARGHRYKHJQSM-UHFFFAOYSA-N
Synonym phenylcyclohexane, benzene, cyclohexyl, cyclohexane, phenyl, cyclohexyl-benzene, cyclohexyl benzene, 4-cyclohexylbenzene, santosol 360, phenyl cyclohexane, 1,1'-biphenyl, 1,2,3,4,5,6-hexahydro, wln: l6tj ar
PubChem CID 13229
IUPAC Name cyclohexylbenzene
SMILES C1CCC(CC1)C1=CC=CC=C1

Specifications

Melting Point 5°C
Color Colorless
Density 0.9500g/mL
Boiling Point 238°C to 240°C
Flash Point 98°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula C6H5C6H11
Refractive Index 1.5250 to 1.527
Quantity 500 mL
Beilstein 05, 503
Specific Gravity 0.95
Solubility Information Solubility in water: insoluble
Formula Weight 160.26
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclohexylbenzene, 98%
Afficher plus Afficher moins

RUO – Research Use Only

Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.