missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexyldiphenylphosphine, 98%

Product Code. 10490821 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10490821 5 g Glass bottle 5g
10278903 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10490821 Supplier Thermo Scientific Acros Supplier No. 406100050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 6372-42-5 | C18H21P | 268.34 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6372-42-5
Molecular Formula C18H21P
Molecular Weight (g/mol) 268.34
MDL Number MFCD00046360
InChI Key ZXKWUYWWVSKKQZ-UHFFFAOYSA-N
Synonym cyclohexyldiphenylphosphine, phosphine, cyclohexyldiphenyl, cyclohexyl diphenyl phosphine, diphenylcyclohexylphosphine, diphenylphosphinocyclohexane, cyclohexyl-diphenyl-phosphane, pubchem6451, acmc-1ba4g, cyclohexyl diphenyl phosphane
PubChem CID 80756
IUPAC Name cyclohexyl(diphenyl)phosphane
SMILES C1CCC(CC1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 53.0°C
Color White
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Solubility Information Solubility in water: insoluble in water
Formula Weight 268.33
Percent Purity 98%
Physical Form Powder
Chemical Name or Material Cyclohexyldiphenylphosphine
Show More Show Less
compliance-icons
Product Identifier
  • Cyclohexyldiphenylphosphine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.