missing translation for 'onlineSavingsMsg'
Learn More

Cyclohexylmethyl isothiocyanate, 97%

Product Code. 11358245
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11358245 5 g 5g
11348245 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11358245 Supplier Thermo Scientific Alfa Aesar Supplier No. L10129.06
 Restricted Item
Estimated Shipment: 02-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 52395-66-1 | C8H13NS | 155.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52395-66-1
Molecular Formula C8H13NS
Molecular Weight (g/mol) 155.26
MDL Number MFCD00040881
InChI Key RYBJNWIQNLZNPE-UHFFFAOYSA-N
Synonym cyclohexanemethyl isothiocyanate, isothiocyanatomethyl cyclohexane, cyclohexylmethyl isothiocyanate, cyclohexylmethanisothiocyanate, cyclohexanemethylisothiocyanate, isothiocyanatomethyl cyclohexane #
PubChem CID 142928
IUPAC Name isothiocyanatomethylcyclohexane
SMILES S=C=NCC1CCCCC1

Specifications

Density 1.02
Boiling Point 222°C to 223°C
Flash Point >110°C (230°F)
Refractive Index 1.536
Quantity 5 g
UN Number UN2922
Beilstein 2079123
Sensitivity Moisture sensitive
Formula Weight 155.26
Percent Purity 97%
Chemical Name or Material Cyclohexylmethyl isothiocyanate
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.