missing translation for 'onlineSavingsMsg'
Learn More

(Cyclohexylmethyl)zinc bromide, 0.5M in THF, packaged under Argon in resealable ChemSeal™ bottles

Product Code. 15435335
Change view
Click to view available options
Quantity:
50 mL
Unit Size:
50mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15435335 50 mL 50mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15435335 Supplier Thermo Scientific Alfa Aesar Supplier No. H58186.AD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 135579-86-1 | C7H13BrZn | 242.465 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 135579-86-1
Density 0.972
Molecular Formula C7H13BrZn
MDL Number MFCD01311418
Quantity 50 mL
UN Number UN3399
Sensitivity Air sensitive
Synonym cyclohexylmethyl zinc bromide solution, cyclohexyl methylzinc bromide, cyclohexylmethyl zinc ii bromide, bromozinc 1+ ,methanidylcyclohexane, cyclohexylmethyl zinc bromide, 0.50 m in thf, cyclohexyl methylzinc bromide 0.5 m in tetrahydrofuran, cyclohexylmethyl zinc bromide solution, 0.5 m in thf, cyclohexylmethyl zinc bromide, 0.5m in thf, packaged under argon in resealable chemseal bottles
InChI Key SJCALMUTSZRVSS-UHFFFAOYSA-M
SMILES [CH2-]C1CCCCC1.[Zn+]Br
IUPAC Name bromozinc(1+);methanidylcyclohexane
Molecular Weight (g/mol) 242.465
PubChem CID 4486981
Formula Weight 242.46
Chemical Name or Material (Cyclohexylmethyl)zinc bromide, Packaged under Argon in resealable ChemSeal™ bottles
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.