missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclohexylmethyldichlorosilane, 97+%, Thermo Scientific Chemicals
Shop All Thermo Scientific Chemicals ProductsDescription
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Colorless to Brown |
| Density | 1.0950g/mL |
| Boiling Point | 202.0°C |
| Flash Point | 66°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 97% min. (GC) |
| Packaging | Glass bottle |
| Linear Formula | C6H11Si(CH3)Cl2 |
| Refractive Index | 1.4710 to 1.4730 (20°C, 589nm) |
| Quantity | 10 mL |
| Show More |
Safety and Handling
Product Identifier
- Cyclohexylmethyldichlorosilane
- Danger
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 1 A
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H314-Causes severe skin burns and eye damage.
- EUH014-Reacts violently with water.
- EUH071-Corrosive to the respiratory tract.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?