missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctanemethanol, 97%, Thermo Scientific Chemicals

Product Code. 11464417
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11464417 5 g 5g
11474417 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11464417 Supplier Thermo Scientific Alfa Aesar Supplier No. B20450.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Cyclooctanemethanol is used as a high boiling solvent and heat transfer agent. It is also useful for conversion into plastic and lacquer base materials. Cyclooctanemethanol can be oxidized to cyclooctane carboxylic acid and the acid can be converted into octahydro-2-oxo-1H-azonine (S-aminooctanoic acid lactam), which is a starting material for the production of nylon-8.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclooctanemethanol is used as a high boiling solvent and heat transfer agent. It is also useful for conversion into plastic and lacquer base materials. Cyclooctanemethanol can be oxidized to cyclooctane carboxylic acid and the acid can be converted into octahydro-2-oxo-1H-azonine (S-aminooctanoic acid lactam), which is a starting material for the production of nylon-8.

Solubility
It is soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under normal temperatures and pressures. Keep away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3637-63-6
Molecular Formula C9H18O
Molecular Weight (g/mol) 142.242
MDL Number MFCD00001749
InChI Key ZHPBLHYKDKSZCQ-UHFFFAOYSA-N
Synonym cyclooctanemethanol, cyclo octyl carbinol, cyclooctylmethan-1-ol, cyclooctylcarbinol, cyclooctane methanol, acmc-20alcc, hydroxymethylcyclooctane
PubChem CID 77196
IUPAC Name cyclooctylmethanol
SMILES C1CCCC(CCC1)CO

Specifications

Density 0.94
Flash Point 93°C (199°F)
Quantity 5 g
Solubility Information It is soluble in water.
Formula Weight 142.24
Percent Purity 97%
Chemical Name or Material Cyclooctanemethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.