missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctanol, 97%

Product Code. 11433418
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433418 25 g 25g
11443418 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11433418 Supplier Thermo Scientific Alfa Aesar Supplier No. A17825.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 696-71-9 | C8H16O | 128.22 g/mol

Cyclooctanol is a key starting material used for the preparation of cylooctanone by using a chromic acid reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclooctanol is a key starting material used for the preparation of cylooctanone by using a chromic acid reagent.

Solubility
Miscible with acetone.

Notes
Contains 75+% cyclopentane; remainder consists of C{5} and C{6} hydrocarbon.Incompatible with strong oxidizing agents, acid chloride, and acid anhydrides.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 696-71-9
Molecular Formula C8H16O
Molecular Weight (g/mol) 128.22
MDL Number MFCD00001744
InChI Key FHADSMKORVFYOS-UHFFFAOYSA-N
Synonym cyclooctyl alcohol, 1-hydroxycyclooctane, acmc-209oac, fhadsmkorvfyos-uhfffaoysa
PubChem CID 12766
IUPAC Name cyclooctanol
SMILES OC1CCCCCCC1

Specifications

Melting Point 14°C to 15°C
Density 0.97
Boiling Point 97°C to 98°C (11 mmHg)
Flash Point 86°C (186°F)
Refractive Index 1.486
Quantity 25 g
Beilstein 1848166
Solubility Information Miscible with acetone.
Formula Weight 128.22
Percent Purity 97%
Chemical Name or Material Cyclooctanol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.