missing translation for 'onlineSavingsMsg'
Learn More

Cyclooctanone oxime, 98+%

Product Code. 11472658
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11472658 50 g 50g
11462658 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11472658 Supplier Thermo Scientific Alfa Aesar Supplier No. A16672.18
 Restricted Item
Estimated Shipment: 24-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 1074-51-7 | C8H15NO | 141.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1074-51-7
Molecular Formula C8H15NO
Molecular Weight (g/mol) 141.21
MDL Number MFCD00191967
InChI Key KTPUHSVFNHULJH-UHFFFAOYSA-N
Synonym cyclooctanone oxime, cyclooctanone, oxime, cyclooctanonoxim, cyclooctanoneoxime, acmc-1bxvz, hydroxyimino cyclooctane
PubChem CID 136841
IUPAC Name N-cyclooctylidenehydroxylamine
SMILES ON=C1CCCCCCC1

Specifications

Melting Point 36°C to 37°C
Boiling Point 128°C to 129°C (14 mmHg)
Quantity 50 g
Formula Weight 141.22
Percent Purity ≥98%
Chemical Name or Material Cyclooctanone oxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.