missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentadienyltitanium trichloride, 99%

Product Code. 10684902 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 g
10 g
Packaging:
Glass bottle
Unit Size:
10g
2.5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10684902 2.5 g Glass bottle 2.5g
10062690 10 g Glass bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10684902 Supplier Thermo Scientific Acros Supplier No. 215770025
 Restricted Item
Estimated Shipment: 05-11-2026
Add to basket
This item is not returnable. View return policy

CAS: 1270-98-0 | C5H5Cl3Ti | 219.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1270-98-0
Molecular Formula C5H5Cl3Ti
Molecular Weight (g/mol) 219.36
MDL Number MFCD00003724
InChI Key QOXHZZQZTIGPEV-UHFFFAOYSA-K

Specifications

Melting Point 170°C
Color Yellow to Orange
Physical Form Crystalline Powder and/or Crystals
Quantity 2.5 g
Assay Percent Range 99%
Solubility Information Solubility in water: reacts
Formula Weight 219.36
Percent Purity 99%
Infrared Spectrum Authentic
Packaging Glass bottle
Chemical Name or Material Cyclopentadienyltitanium trichloride, 99%
Show More Show Less
compliance-icons
Product Identifier
  • Cyclopentadienyltitanium trichloride
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.