missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentane, 95+%, Extra Dry, AcroSeal™

Product Code. 13128493 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
13128493 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13128493 Supplier Thermo Scientific Acros Supplier No. 397680010
  • ONLINE EXCLUSIVE PRICE
    122.00 €
    118.00 € / 1 L
    Save 4.00 €
    3% Off
 Restricted Item
Estimated Shipment: 21-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 287-92-3 | C5H10 | 70.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 287-92-3
Melting Point -94.0°C
Color Colorless
Density 0.7500g/mL
Boiling Point 50.0°C
Flash Point −7°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Molecular Formula C5H10
Linear Formula C5H10
Refractive Index 1.4050 to 1.4070
MDL Number MFCD00001356
Quantity 1 L
Residue after Evaporation 0.001% max.
Merck Index 15, 2736
Specific Gravity 0.75
Synonym pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in acetone,benzene,alcohol,ether and,most hydrocarbon solvents
InChI Key RGSFGYAAUTVSQA-UHFFFAOYSA-N
SMILES C1CCCC1
IUPAC Name cyclopentane
Molecular Weight (g/mol) 70.15
PubChem CID 9253
ChEBI CHEBI:23492
Viscosity 0.44 mPa.s (20°C)
Formula Weight 70.15
Percent Purity 95+%
Grade Extra Dry
Physical Form Liquid
Chemical Name or Material Cyclopentane, Extra Dry
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.