missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentane, CHROMASOLV™, for HPLC, ≥75% cyclopentane basis, Solstice

Product Code. 15632860
Change view
Click to view available options
Quantity:
1 L
Packaging:
Glass Bottle
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15632860 1 L Glass Bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15632860 Supplier Honeywell-Chromasolv Supplier No. 2706011L
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CHROMASOLV™, for HPLC,≥75% cyclopentane basis

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Specifications

CAS 287-92-3
Absorbance max. 0.01 at 400 nm, max. 0.01 at 240 nm, max. 0.10 at 220 nm, max. 0.50 at 210 nm, max. 1.0 at 198 nm
CAS Max % 100.0000%
Melting Point -94°C (literature)
Density 0.751 g/mL at 25 °C(lit.)
Boiling Point 50°C (literature)
Flash Point −20°C
Packaging Glass Bottle
Molecular Formula C5H10
Linear Formula C5H10
MDL Number MFCD00001356
Quantity 1 L
UN Number UN1146
Beilstein 1900195
Synonym pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
InChI Key RGSFGYAAUTVSQA-UHFFFAOYSA-N
SMILES C1CCCC1
IUPAC Name cyclopentane
Molecular Weight (g/mol) 70.135
PubChem CID 9253
ChEBI CHEBI:23492
Formula Weight 70.13
Percent Purity >75%
Grade HPLC
Chemical Name or Material Cyclopentane, CHROMASOLV™, for HPLC, ≥75% cyclopentane basis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.