missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentane, HPLC Grade

Product Code. 11480101
Change view
Click to view available options
Quantity:
250 mL
1 L
4 L
Unit Size:
1 L
250mL
4 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11480101 1 L 1 L
11470101 250 mL 250mL
11490101 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11480101 Supplier Thermo Scientific Alfa Aesar Supplier No. 044814.K2
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For HPLC and Spectrophotometry | CAS: 287-92-3 | C5H10 | 70.135 g/mol

Cyclopentane is used in all analytical laboratories as a major component of mobile phase in High Performance Liquid Chromatography (HPLC) and in Liquid Chromatography - Mass Spectrometry (LC-MS).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 287-92-3
Molecular Formula C5H10
Molecular Weight (g/mol) 70.135
MDL Number MFCD00001356
InChI Key RGSFGYAAUTVSQA-UHFFFAOYSA-N
Synonym pentamethylene, ciclopentano, cyclopentan, zyklopentan, unii-t86pb90rnu, hsdb 62, t86pb90rnu, cyclopentanes, cyclopentyl group, hydrocarbons, cyclic c5 and c6
PubChem CID 9253
ChEBI CHEBI:23492
IUPAC Name cyclopentane
SMILES C1CCCC1

Specifications

Melting Point °C to 95°C
Color Colorless
Density 0.748
Boiling Point 47°C to 49°C
Flash Point -37°C (-34°F)
Ignition Point 380°C (716°F)
Odor Petrol-like
Refractive Index 1.406
Quantity 1 L
Beilstein 1900195
Solubility Information Miscible with ethanol,ether and acetone. Slightly miscible with water.
Formula Weight 70.14
Vapor Pressure 398mm Hg at 20°C
Grade HPLC
Physical Form Liquid
Chemical Name or Material Cyclopentane, Contains 75+% cyclopentane. Remainder consists of C5 and C6 hydrocarbons .
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H336-May cause drowsiness or dizziness.
  • H412-Harmful to aquatic life with long lasting effects.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.