missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanesulfonyl chloride, 97%

Product Code. 15485627
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15485627 250 mg 250mg
15465627 1 g 1g
15475627 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15485627 Supplier Thermo Scientific Alfa Aesar Supplier No. H32865.MD
 Restricted Item
Estimated Shipment: 16-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 26394-17-2 | C5H9ClO2S | 168.635 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26394-17-2
Molecular Formula C5H9ClO2S
Molecular Weight (g/mol) 168.635
MDL Number MFCD04117774
InChI Key HZVKYZHPDGEECE-UHFFFAOYSA-N
Synonym cyclopentylsulfonyl chloride, cyclopentanesulphonyl chloride, chlorocyclopentylsulfone, cyclopentylsulfonylchloride, acmc-1ccr5, cyclopentanesulfonylchloride, cyclopentyl sulfonyl chloride, chlorosulphonyl cyclopentane, cyclopentane sulfonyl chloride, cyclopentane sulphonyl chloride
PubChem CID 3717197
IUPAC Name cyclopentanesulfonyl chloride
SMILES C1CCC(C1)S(=O)(=O)Cl

Specifications

Boiling Point 71°C to 73°C (2 mmHg)
Quantity 250 mg
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 168.64
Percent Purity 97%
Chemical Name or Material Cyclopentanesulfonyl chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.