missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanethiol, 98%

Product Code. 10361190 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10361190

Brand: Thermo Scientific Acros 435690250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1679-07-8 | C5H10S | 102.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1679-07-8
Molecular Formula C5H10S
Molecular Weight (g/mol) 102.20
MDL Number MFCD00001369
InChI Key WVDYBOADDMMFIY-UHFFFAOYSA-N
Synonym cyclopentyl mercaptan, mercaptocyclopentane, cyclopentylthiol, fema no. 3262, cyclopentyl thiol, cyclopen-tanethiol, cyclopentane thiol, cyclopentylmercaptan, acmc-209dxb, cyclopentanethiol
PubChem CID 15510
IUPAC Name cyclopentanethiol
SMILES SC1CCCC1

Specifications

Color Colorless to Yellow
Boiling Point 129°C to 131°C
Flash Point 25°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Quantity 25 g
Formula Weight 102.2
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclopentanethiol
compliance-icons compliance-icons
Product Identifier
  • Cyclopentanethiol
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.