missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentanone-2,2,5,5-d4, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | Cyclopentanone-2,2,5,5-D4 |
| CAS | 3997-89-5 |
| Molecular Formula | C5 D4 H4 O |
| Quantity | 1 g |
| Synonym | Cyclopentanone-2,2,5,5-d4 (6 CI, 8 CI, 9 CI, ACI), 2,2,5,5-Tetradeuteriocyclopentan-1-one, Cyclopentanone-α-d4, Cyclopentanone-2,2,5,5-D4, Cyclopentanone-d4 |
| InChI Formula | InChI=1 S/C5H8O/c6-5-3-1-2-4-5/h1-4H2/i3D2,4D2 |
| SMILES | [2 H]C1([2 H])CCC([2 H])([2 H])C1=O |
| IUPAC Name | 2,2,5,5-tetradeuteriocyclopentan-1-one |
| Molecular Weight (g/mol) | 88.1411 |
| Formula Weight | 88.0826 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction