missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentanone oxime, 97%

Product Code. 11441507
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11441507 100 g 100g
11431507 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11441507 Supplier Thermo Scientific Alfa Aesar Supplier No. B24961.22
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1192-28-5 | C5H9NO | 99.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1192-28-5
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00001420
InChI Key YGNXYFLJZILPEK-UHFFFAOYSA-N
Synonym cyclopentanone oxime, cyclopentanoneoxime, cyclopentanone, oxime, unii-qy3bv00pcy, ccris 7163, qy3bv00pcy, n-1e-cyclopentylidene hydroxylamine, hydroxyimino cyclopentane, cyclopentanone,oxime, hydroxyiminocyclopentane
PubChem CID 14500
IUPAC Name N-cyclopentylidenehydroxylamine
SMILES ON=C1CCCC1

Specifications

Melting Point 54°C to 58°C
Boiling Point 196°C
Flash Point 92°C (198°F)
Quantity 100 g
Formula Weight 99.13
Percent Purity 97%
Chemical Name or Material Cyclopentanone oxime

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.