missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentene-1-boronic acid, 97%

Product Code. 15479267
Change view
Click to view available options
Quantity:
250 mg
Unit Size:
250mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15479267 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15479267 Supplier Thermo Scientific Alfa Aesar Supplier No. H52531.MD
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 850036-28-1 | C5H9BO2 | 111.935 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 850036-28-1
Melting Point 134°C to 140°C
Molecular Formula C5H9BO2
MDL Number MFCD02179496
Quantity 250 mg
Synonym cyclopent-1-en-1-ylboronic acid, 1-cyclopentenylboronic acid, cyclopentenylboronic acid, cyclopent-1-en-1-yl boronic acid, cyclopentene-1-boronic acid, cyclopenten-1-yl boronic acid, cyclopent-1-ene-1-boronic acid, boronic acid, 1-cyclopenten-1-yl, cyclopenteneboronic acid, acmc-209pzb
InChI Key UZBHNSVUMGIKLU-UHFFFAOYSA-N
SMILES B(C1=CCCC1)(O)O
IUPAC Name cyclopenten-1-ylboronic acid
Molecular Weight (g/mol) 111.935
PubChem CID 3861343
Formula Weight 111.94
Percent Purity 97%
Chemical Name or Material Cyclopentene-1-boronic acid
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.