missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentene oxide, 97%

Product Code. 11469057
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11469057 5 g 5g
11479057 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11469057 Supplier Thermo Scientific Alfa Aesar Supplier No. B20629.06
 Restricted Item
Estimated Shipment: 01-09-2026
Add to basket
This item is not returnable. View return policy

CAS: 285-67-6 | C5H8O | 84.12 g/mol

Cyclopentene oxide is used to prepare beta-amino alcohols by reacting with aromatic amines using bismuth trichloride as a catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentene oxide is used to prepare beta-amino alcohols by reacting with aromatic amines using bismuth trichloride as a catalyst.

Solubility
Immiscible with water.

Notes
Store in a cool place. Incompatible with acids, bases and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 285-67-6
Molecular Formula C5H8O
Molecular Weight (g/mol) 84.12
MDL Number MFCD00005161
InChI Key GJEZBVHHZQAEDB-UHFFFAOYNA-N
Synonym cyclopentene oxide, 6-oxabicyclo 3.1.0 hexane, 1,2-epoxycyclopentane, cyclopentane oxide, cyclopentene epoxide, epoxycyclopentane, cis-1,2-epoxycyclopentane, cyclopentane, 1,2-epoxy, 6-oxa-bicyclo 3.1.0 hexane
PubChem CID 9244
IUPAC Name 6-oxabicyclo[3.1.0]hexane
SMILES C1CC2OC2C1

Specifications

Density 0.965
Boiling Point 101°C to 102°C
Flash Point 10°C (50°F)
Refractive Index 1.434
Quantity 5 g
UN Number UN1993
Beilstein 102495
Solubility Information Immiscible with water.
Formula Weight 84.12
Percent Purity 97%
Chemical Name or Material Cyclopentene oxide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.