missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyclopentyl Chloride, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Cyclopentyl Chloride |
| CAS | 930-28-9 |
| Molecular Formula | C5H9Cl |
| Quantity | 50 mg |
| InChI Formula | InChI=1S/C5H9Cl/c6-5-3-1-2-4-5/h5H,1-4H2 |
| SMILES | C1CCC(C1)Cl |
| IUPAC Name | chlorocyclopentane |
| Molecular Weight (g/mol) | 104.58 |
| Formula Weight | 104.0393 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction