missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl iodide, 98%, stabilized over copper

Product Code. 11944511 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11944511 25 mL Glass bottle 25mL
11934511 5 mL Glass bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11944511 Supplier Thermo Scientific Acros Supplier No. 381730250
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1556-18-9 | C5H9I | 196.03 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1556-18-9
Molecular Formula C5H9I
Molecular Weight (g/mol) 196.03
MDL Number MFCD00039446
InChI Key PCEBAZIVZVIQEO-UHFFFAOYSA-N
Synonym cyclopentane, iodo, cyclopentyl iodide, jodcyclopentan, cyclopentyliodide, iodo-cyclopentane, iodocyclo-pentane, iodocyclopentane, acmc-1c0qu
PubChem CID 73784
IUPAC Name iodocyclopentane
SMILES IC1CCCC1

Specifications

Color Colorless to Yellow
Density 1.6950g/mL
Boiling Point 166.0°C to 167.0°C
Flash Point 52°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C5H9I
Quantity 25 mL
Specific Gravity 1.695
Stabilizer (Copper)
Formula Weight 196.03
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material Cyclopentyl iodide, Stabilized
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclopentyl iodide, stabilized over copper
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.