missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl isocyanate, 97%

Product Code. 10684745 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10684745 5 g 5g
10684555 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10684745 Supplier Thermo Scientific Acros Supplier No. 397370050
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 4747-71-1 | C6H9NO | 111.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4747-71-1
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.14
MDL Number MFCD02093695
InChI Key CZALJDQHONFVFU-UHFFFAOYSA-N
Synonym cyclopentyl isocyanate, cyclopentylisocyanate, cyclopentane, isocyanato, isocyanic acid cyclopentyl ester, isocyanato cyclopentane, isocyanato-cyclopentane, acmc-1aspa, ksc497s6f
PubChem CID 643500
IUPAC Name isocyanatocyclopentane
SMILES C1CCC(C1)N=C=O

Specifications

Density 0.9860g/mL
Boiling Point 147.0°C
Flash Point 36°C
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula C5H9NCO
Quantity 5 g
Specific Gravity 0.986
Formula Weight 111.14
Percent Purity 97%
Chemical Name or Material Cyclopentyl isocyanate
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Cyclopentyl isocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.