missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl methyl ether, Reagent Grade, contains 50 ppm BHT as inhibitor, ≥99.90%, Honeywell™

Product Code. 15626690 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
5 L
Packaging:
Aluminum bottle
Glass bottle
Unit Size:
1 L
5 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15626690 500 mL Glass bottle 500mL
15606690 1 L Glass bottle 1 L
15636690 5 L Aluminum bottle 5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15626690 Supplier Honeywell Research Chemicals Supplier No. 675989500ML
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, contains 50 ppm BHT as inhibitor,≥99.90%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5614-37-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD08276401
InChI Key SKTCDJAMAYNROS-UHFFFAOYSA-N
Synonym cyclopentyl methyl ether, cyclopentane, methoxy, cpme, cyclopentane,methoxy, unii-4067e5gbkb, cyclopentylmethylether, cylopentylmethylether, methoxycyclopentane, 2-methoxycyclopentyl, cylopentyl-methylether
PubChem CID 138539
IUPAC Name methoxycyclopentane
SMILES COC1CCCC1

Specifications

Melting Point 140°C
Density 0.86g/mL at 25°C
Boiling Point 106°C (1013 hPa)
Packaging Glass bottle
Quantity 500 mL
UN Number UN3271
Formula Weight 100.16g/mol
Percent Purity ≥99.90%
Chemical Name or Material Cyclopentyl methyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.