missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl methyl ether, Reagent Grade, contains 50 ppm BHT as inhibitor, ≥99.90%, Honeywell™

Product Code. p-7120365 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
500 mL
1 L
5 L
Packaging:
Aluminum bottle
Glass bottle
Unit Size:
1 L
5 L
500mL
Product Code. Quantity Packaging unitSize
15626690 500 mL Glass bottle 500mL
15606690 1 L Glass bottle 1 L
15636690 5 L Aluminum bottle 5 L
3 options
This item is not returnable. View return policy

Product Code. 15626690

Brand: Honeywell Research Chemicals 675989500ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade, contains 50 ppm BHT as inhibitor,≥99.90%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5614-37-9
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD08276401
InChI Key SKTCDJAMAYNROS-UHFFFAOYSA-N
Synonym cyclopentyl methyl ether, cyclopentane, methoxy, cpme, cyclopentane,methoxy, unii-4067e5gbkb, cyclopentylmethylether, cylopentylmethylether, methoxycyclopentane, 2-methoxycyclopentyl, cylopentyl-methylether
PubChem CID 138539
IUPAC Name methoxycyclopentane
SMILES COC1CCCC1

Specifications

Melting Point 140°C
Density 0.86g/mL at 25°C
Boiling Point 106°C (1013 hPa)
Packaging Glass bottle
Quantity 500 mL
UN Number UN3271
Formula Weight 100.16g/mol
Percent Purity ≥99.90%
Chemical Name or Material Cyclopentyl methyl ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.