missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl Methyl Ether, TRC

Product Code. 30357914
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30357914 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30357914 Supplier Toronto Research Chemicals Supplier No. TRCC9923381G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 5614-37-9
Molecular Formula C6H12O
Quantity 1 g
Synonym Ether, cyclopentyl methyl (7CI,8CI), Methoxycyclopentane, CPME, Cyclopentyl methyl ether, Methyl cyclopentyl ether, Methoxycyclopentane, CPME, Methyl Cyclopentyl Ether
InChI Formula InChI=1S/C6H12O/c1-7-6-4-2-3-5-6/h6H,2-5H2,1H3
SMILES COC1CCCC1
IUPAC Name methoxycyclopentane
Molecular Weight (g/mol) 100.16
Formula Weight 100.0888
Chemical Name or Material Cyclopentyl Methyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.