missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentyl phenyl ketone, 96%

Product Code. 11438857
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11438857 5 g 5g
11428857 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11438857 Supplier Thermo Scientific Alfa Aesar Supplier No. A19976.06
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 5422-88-8 | C12H14O | 174.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5422-88-8
Molecular Formula C12H14O
Molecular Weight (g/mol) 174.24
MDL Number MFCD00001378
InChI Key VYDIMQRLNMMJBW-UHFFFAOYSA-N
Synonym cyclopentyl phenyl ketone, cyclopentyl phenyl methanone, benzoylcyclopentane, methanone, cyclopentylphenyl, ketone, cyclopentyl phenyl, cyclopentyl-phenyl-methanone, cyclopentylphenylketone, acmc-209leg, phenyl cyclopentyl ketone, cyclopentanylphenylmethanone
PubChem CID 79464
IUPAC Name cyclopentyl(phenyl)methanone
SMILES O=C(C1CCCC1)C1=CC=CC=C1

Specifications

Density 1.036
Boiling Point 136°C to 140°C (16 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.544
Quantity 5 g
Beilstein 2045903
Formula Weight 174.24
Percent Purity 96%
Chemical Name or Material Cyclopentyl phenyl ketone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.