missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylacetic acid, 97%

Product Code. 10734271 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
Packaging:
Glass bottle
Unit Size:
10g
25g
This item is not returnable. View return policy

Product Code. 10734271

Brand: Thermo Scientific Acros 154870250

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 1123-00-8 | C7H12O2 | 128.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1123-00-8
Molecular Formula C7H12O2
Molecular Weight (g/mol) 128.17
MDL Number MFCD00001387
InChI Key YVHAIVPPUIZFBA-UHFFFAOYSA-N
Synonym cyclopentylacetic acid, cyclopentaneacetic acid, cycylopentylacetic acid, carboxymethyl cyclopentane, cyclopentyl acetic acid, cyclopentyacetic acid, cyclopentanacetic acid, cyclopentyl-acetic acid, 2-cyclopentylaceticacid, cyclopentane acetic acid
PubChem CID 71606
IUPAC Name 2-cyclopentylacetic acid
SMILES OC(=O)CC1CCCC1

Specifications

Color Colorless
Density 1.0200g/mL
Boiling Point 133.0°C to 134.0°C (23.0 mmHg)
Flash Point 109°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula C5H9CH2CO2H
Refractive Index 1.4520 to 1.4540
Quantity 25 g
Beilstein 09, 10
Specific Gravity 1.02
Formula Weight 128.17
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material Cyclopentylacetic acid, 97%
Show More Show Less
compliance-icons
Product Identifier
  • Cyclopentylacetic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.