missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylacetylene, 97%

Product Code. 15480348
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15480348

Brand: Thermo Scientific Alfa Aesar H53483.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 930-51-8 | C7H10 | 94.16 g/mol

Cyclopentylacetylene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentylacetylene is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-51-8
Molecular Formula C7H10
Molecular Weight (g/mol) 94.16
MDL Number MFCD00013744
InChI Key TXVJSWLZYQMWPC-UHFFFAOYSA-N
Synonym cyclopentylacetylene, cyclopentyl acetylene, cyclopentylethyne, cyclopentane, ethynyl, cyclopentane, ethynyl-7ci, 8ci, 9ci, ethynylcylopentane, ethynyl-cyclopentane, cyclopentyl-acetylen
PubChem CID 136725
IUPAC Name ethynylcyclopentane
SMILES C#CC1CCCC1

Specifications

Density 0.812
Boiling Point 107°C to 109°C
Flash Point 1°C (34°F)
Refractive Index 1.432
Quantity 1 g
UN Number UN3295
Formula Weight 94.16
Percent Purity 97%
Chemical Name or Material Cyclopentylacetylene
compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H225-Highly flammable liquid and vapour.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.