missing translation for 'onlineSavingsMsg'
Learn More

Cyclopentylacetylene, 97%

Product Code. 15490348
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15490348 5 g 5g
15480348 1 g 1g
15400358 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490348 Supplier Thermo Scientific Alfa Aesar Supplier No. H53483.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 930-51-8 | C7H10 | 94.16 g/mol

Cyclopentylacetylene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Cyclopentylacetylene is used as pharmaceutical intermediate.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 930-51-8
Molecular Formula C7H10
Molecular Weight (g/mol) 94.16
MDL Number MFCD00013744
InChI Key TXVJSWLZYQMWPC-UHFFFAOYSA-N
Synonym cyclopentylacetylene, cyclopentyl acetylene, cyclopentylethyne, cyclopentane, ethynyl, cyclopentane, ethynyl-7ci, 8ci, 9ci, ethynylcylopentane, ethynyl-cyclopentane, cyclopentyl-acetylen
PubChem CID 136725
IUPAC Name ethynylcyclopentane
SMILES C#CC1CCCC1

Specifications

Density 0.812
Boiling Point 107°C to 109°C
Flash Point 1°C (34°F)
Refractive Index 1.432
Quantity 5 g
UN Number UN3295
Formula Weight 94.16
Percent Purity 97%
Chemical Name or Material Cyclopentylacetylene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.